missing translation for 'onlineSavingsMsg'
Learn More

(6-Chloro-2-pyridyl)hydrazine, 95%

Product Code. 11342269
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11342269 1 g 1g
15423647 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11342269 Supplier Thermo Scientific Alfa Aesar Supplier No. H27249.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 5193-03-3 | C9H13NO | 151.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5193-03-3
MDL Number MFCD00214724

Specifications

Boiling Point 117°C to 120°C
Molecular Formula C9H13NO
Quantity 1 g
Synonym 5-ethyl-2-pyridineethanol, 2-5-ethylpyridin-2-yl ethanol, 2-5-ethyl-2-pyridyl ethanol, 5-ethyl-2-2-hydroxyethyl pyridine, 5-ethyl-2-hydroxyethylpyridine, 2-5-ethyl-2-pyridinyl-1-ethanol, 2-pyridineethanol, 5-ethyl, 2-hydroxyethyl-5-ethylpyridine, 5-ethylpyridine-2-ethanol, unii-o6f6rcv39p
InChI Key OUJMXIPHUCDRAS-UHFFFAOYSA-N
SMILES CCC1=CN=C(C=C1)CCO
IUPAC Name 2-(5-ethylpyridin-2-yl)ethanol
Molecular Weight (g/mol) 151.21
PubChem CID 78894
Formula Weight 143.58
Percent Purity 95%
Chemical Name or Material (6-Chloro-2-pyridyl)hydrazine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.