missing translation for 'onlineSavingsMsg'
Learn More

6-Methylpyridine-2-carboxylic acid, 95%

Product Code. 11411160
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11411160 1 g 1g
11421160 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11411160 Supplier Thermo Scientific Alfa Aesar Supplier No. H26302.03
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 934-60-1 | C7H7NO2 | 137.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 934-60-1
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.14
MDL Number MFCD00023481
InChI Key LTUUGSGSUZRPRV-UHFFFAOYSA-N
Synonym 6-methyl-2-pyridinecarboxylic acid, 6-methylpicolinic acid, 6-methyl-2-picolinic acid, 2-carboxy-6-methylpyridine, 6-methyl-2-picolinicacid, 2-pyridinecarboxylic acid, 6-methyl, 2-picoline-6-carboxylic acid, picolinic acid, 6-methyl, 6-methyl picolinic acid, unii-kb675y352l
PubChem CID 70282
IUPAC Name 6-methylpyridine-2-carboxylic acid
SMILES CC1=CC=CC(=N1)C(O)=O

Specifications

Melting Point 127°C to 131°C
Boiling Point 100°C (4.5 mmHg)
Odor Odorless
Quantity 1 g
Formula Weight 137.14
Percent Purity 95%
Chemical Name or Material 6-Methylpyridine-2-carboxylic acid
compliance-icons
Product Identifier
  • 6-Methylpyridine-2-carboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 6-Methylpyridine-2-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.