missing translation for 'onlineSavingsMsg'
Learn More

6-Nitropiperonyl Alcohol, TRC

Product Code. 30179251
Change view
Click to view available options
Quantity:
25 mg
50 mg
250 mg
Unit Size:
250mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179251 250 mg 250mg
30179455 25 mg 25mg
30179602 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30179251 Supplier Toronto Research Chemicals Supplier No. TRCN900783250MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15341-08-9
Molecular Formula C8H7NO5
Molecular Weight (g/mol) 197.14
IUPAC Name (6-nitro-1,3-benzodioxol-5-yl)methanol
SMILES OCc1cc2OCOc2cc1[N+](=O)[O-]

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C8H7NO5/c10-3-5-1-7-8(14-4-13-7)2-6(5)9(11)12/h1-2,10H,3-4H2
Formula Weight 197.0324
Chemical Name or Material 6-Nitropiperonyl Alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.