Learn More
7-Bromo-4-methyl-3,4-dihydro-2H-1,4-benzoxazine, 95%, Thermo Scientific™
Product Code. 10662243
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10662243 10 g Amber Glass Bottle 10g
10732083 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10662243 Supplier Thermo Scientific Maybridge Supplier No. CC13510EA
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
missing translation for 'validMainframeMsg'
Chemical Identifiers
CAS 154264-95-6
Molecular Formula C9H10BrNO
Molecular Weight (g/mol) 228.09
MDL Number MFCD02681913
InChI Key MQMFOFZKZBLSAB-UHFFFAOYSA-N
Synonym 7-bromo-4-methyl-3,4-dihydro-2h-1,4-benzoxazine, 7-bromo-3,4-dihydro-4-methyl-2h-1,4-benzoxazine, 2h-1,4-benzoxazine, 7-bromo-3,4-dihydro-4-methyl, 7-bromo-4-methyl-3,4-dihydro-2h-benzo b 1,4 oxazine, 7-bromo-4-methyl-2h,3h-benzo e 1,4-oxazine, 2h-1,4-benzoxazine,7-bromo-3,4-dihydro-4-methyl, 7-bromo-4-methyl-3,4-dihydro-2h-benzo 1,4 oxazine
PubChem CID 2776405
IUPAC Name 7-bromo-4-methyl-2,3-dihydro-1,4-benzoxazine
SMILES CN1CCOC2=C1C=CC(Br)=C2
Specifications
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 228.09
Percent Purity 95%
Chemical Name or Material 7-Bromo-4-methyl-3, 4-dihydro-2H-1, 4-benzoxazine