missing translation for 'onlineSavingsMsg'
Learn More

7-Hydroxy Coumarin Beta-D-Glucuronide Sodium Salt, TRC

Product Code. 17909544
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17909544 5 mg 5mg
17989154 1 mg 1mg
17968674 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17909544 Supplier Toronto Research Chemicals Supplier No. TRCH9248805MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 168286-98-4
Molecular Formula C15 H13 O9 . Na
Molecular Weight (g/mol) 360.25
Synonym 7-Hydroxy Coumarin-B-D-Glucuronide Sodium Salt
IUPAC Name sodium;(2S,3S,4S,5R,6S)-3,4,5-trihydroxy-6-(2-oxochromen-7-yl)oxyoxane-2-carboxylate
SMILES [Na+].O[C@H]1[C@H](Oc2ccc3C=CC(=O)Oc3c2)O[C@@H]([C@@H](O)[C@@H]1O)C(=O)[O-]

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C15H14O9.Na/c16-9-4-2-6-1-3-7(5-8(6)23-9)22-15-12(19)10(17)11(18)13(24-15)14(20)21;/h1-5,10-13,15,17-19H,(H,20,21);/q;+1/p-1/t10-,11-,12+,13-,15+;/m0./s1
Formula Weight 360.0457
Percent Purity >95%
Chemical Name or Material 7-Hydroxycoumarin beta-D-glucuronide sodium salt
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.