missing translation for 'onlineSavingsMsg'
Learn More

Abcam Anti-ATM antibody [EPR20100] - BSA and Azide free, 100 ug

Product Code. 32164647 Shop All Abcam Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32164647 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32164647 Supplier Abcam Supplier No. ab223533100ug
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Monoclonal antibody, suitable for IP, ChIP, WB, ICC/IF, with 1 citations.

  • Host Species: Rabbit
  • Clonality: Monoclonal
  • Isotype: IgG
  • Applications: IP, ChIP, WB, ICC/IF, Flow Cyt (Intra)
  • Species Reactivity: Mouse, Rat and Human
  • Storage Buffer: pH: 7.2 - 7.4, Constituents: PBS
  • Form: Liquid
  • Purification: Affinity purification Protein A

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Antigen ATM
Applications Immunoprecipitation, ChIP sequencing (ChIP-seq), Western Blot, Immunocytochemistry, Immunofluorescence, Flow Cytometry (Intracellular Staining)
Classification Monoclonal
Clone EPR20100
Conjugate Unconjugated
Formulation pH: 7.2 - 7.4, Constituents: PBS
Gene ATM
Gene Symbols ATM
Host Species Rabbit
Purification Method Affinity purification Protein A
Quantity 100 μg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Packaging Vial
Test Specificity This product reacts with Mouse, Rat and Human.
Target Species Mouse, Rat, Human
Product Type Monoclonal Antibody (ATM)
Form Liquid
Isotype IgG
Host abrv Rb
Shipping Condition Blue Ice
Reactivity abrv Ms, Rt, Hu
Target Gene ATM
Target Entry ATM
Target Molecule ATM, Mouse, Rat, Human
Technique IP, ChIP, WB, ICC/IF, Flow Cyt (Intra)
Validated Application abrv IP, ChIP, WB, ICC/IF, Flow Cyt (Intra)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.