missing translation for 'onlineSavingsMsg'
Learn More

Abcam Anti-gamma H2A.X (phospho S139) antibody [EP854(2)Y], 100 uL

Product Code. 32171068 Shop All Abcam Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32171068 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32171068 Supplier Abcam Supplier No. ab81299100uL
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Monoclonal antibody, suitable for IP, Dot, WB, IHC-P, with over 400 citations.

  • Host Species: Rabbit
  • Clonality: Monoclonal
  • Isotype: IgG
  • Applications: IP, Dot, WB, IHC-P, ICC/IF
  • Species Reactivity: Mouse, Rat and Human
  • Storage Buffer: pH: 7.2 - 7.4, Preservative: 0.01% Sodium azide, Constituents: PBS, 40% Glycerol (glycerin, glycerine), 0.05% BSA
  • Form: Liquid
  • Purification: Affinity purification Protein A

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Antigen H2AX
Applications Immunoprecipitation, Dot Blot, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone EP854(2)Y
Conjugate Unconjugated
Formulation pH: 7.2 - 7.4, Preservative: 0.01% Sodium azide, Constituents: PBS, 40% Glycerol (glycerin, glycerine), 0.05% BSA
Gene H2AX
Gene Symbols H2AX
Host Species Rabbit
Purification Method Affinity purification Protein A
Quantity 100 μL
Regulatory Status RUO
Chains or Fragments IgG
Whole Molecule Yes
Primary or Secondary Primary
Packaging Vial
Test Specificity This product reacts with Mouse, Rat and Human.
Target Species Mouse, Rat, Human
Product Type Monoclonal Antibody (H2AX)
Form Liquid
Format Vial
Isotype IgG
Host abrv Rb
Shipping Condition Blue Ice
Reactivity abrv Ms, Rt, Hu
Target Modification State phospho S139
Target Gene H2AX
Target Entry H2AX
Target Molecule H2AX, Mouse, Rat, Human
Technique IP, Dot, WB, IHC-P, ICC/IF
Validated Application abrv IP, Dot, WB, IHC-P, ICC/IF
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.