missing translation for 'onlineSavingsMsg'
Learn More

Abcam Anti-GFP antibody, 100 ug

Product Code. 32103973 Shop All Abcam Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32103973 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32103973 Supplier Abcam Supplier No. ab6673100ug
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Goat Polyclonal antibody, suitable for IP, ELISA, WB, IHC-P, with over 600 citations.

  • Host Species: Goat
  • Clonality: Polyclonal
  • Isotype: IgG
  • Applications: IP, ELISA, WB, IHC-P, ICC/IF, IHC-Fr, IHC - Wmt
  • Storage Buffer: pH: 7.2, Preservative: 0.01% Sodium azide, Constituents: 0.88% Sodium chloride, 0.424% Potassium phosphate solution
  • Form: Liquid
  • Purification: Affinity purification

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Antigen GFP
Applications Immunoprecipitation, ELISA, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Whole-Mount)
Classification Polyclonal
Conjugate Unconjugated
Formulation pH: 7.2, Preservative: 0.01% Sodium azide, Constituents: 0.88% Sodium chloride, 0.424% Potassium phosphate solution
Gene GFP
Gene Symbols GFP
Host Species Goat
Purification Method Affinity purification
Quantity 100 μg
Regulatory Status RUO
Chains or Fragments IgG
Whole Molecule Yes
Primary or Secondary Primary
Packaging Vial
Target Species Non-species specific
Product Type Polyclonal Antibody (GFP)
Form Liquid
Format Vial
Isotype IgG
Host abrv Gt
Shipping Condition Blue Ice
Target Gene GFP
Target Entry GFP
Target Molecule GFP
Technique IP, ELISA, WB, IHC-P, ICC/IF, IHC-Fr, IHC - Wmt
Validated Application abrv IP, ELISA, WB, IHC-P, ICC/IF, IHC-Fr, IHC - Wmt
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.