missing translation for 'onlineSavingsMsg'
Learn More

Abcam Anti-KAP1 (phospho S824) antibody [BL-246-7B5], 100 uL

Product Code. 32094807 Shop All Abcam Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32094807 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32094807 Supplier Abcam Supplier No. ab243870100uL
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Monoclonal antibody, suitable for IHC-P, ICC/IF, IP, WB, with 3 citations.

  • Host Species: Rabbit
  • Clonality: Monoclonal
  • Isotype: IgG
  • Applications: IHC-P, ICC/IF, IP, WB, Flow Cyt
  • Species Reactivity: Mouse and Human
  • Storage Buffer: pH: 7.8 - 8.6, Preservative: 0.09% Sodium azide, Constituents: 98% Borate buffered saline, 0.1% BSA
  • Form: Liquid
  • Purification: Affinity purification Protein A

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Antigen TRIM28
Applications Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot, Flow Cytometry
Classification Monoclonal
Clone BL-246-7B5
Conjugate Unconjugated
Formulation pH: 7.8 - 8.6, Preservative: 0.09% Sodium azide, Constituents: 98% Borate buffered saline, 0.1% BSA
Gene TRIM28
Gene Symbols TRIM28
Host Species Rabbit
Purification Method Affinity purification Protein A
Quantity 100 μL
Regulatory Status RUO
Chains or Fragments IgG
Whole Molecule Yes
Primary or Secondary Primary
Packaging Vial
Test Specificity This product reacts with Mouse and Human.
Target Species Mouse, Human
Product Type Monoclonal Antibody (TRIM28)
Form Liquid
Format Vial
Isotype IgG
Host abrv Rb
Shipping Condition Blue Ice
Reactivity abrv Ms, Hu
Target Modification State phospho S824
Target Gene TRIM28
Target Entry TRIM28
Target Molecule TRIM28, Mouse, Human
Technique IHC-P, ICC/IF, IP, WB, Flow Cyt
Validated Application abrv IHC-P, ICC/IF, IP, WB, Flow Cyt
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.