missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABCB8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-59553
This item is not returnable.
View return policy
Description
ABCB8 Polyclonal specifically detects ABCB8 in Human samples. It is validated for Western Blot.
Specifications
| ABCB8 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| ATP-binding cassette, sub-family B (MDR/TAP), member 8, EC 3.6.3, EST328128, M-ABC1mitochondrial ABC protein, mitochondrial, Mitochondrial ATP-binding cassette 1 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 11194 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q9NUT2 | |
| ABCB8 | |
| Synthetic peptides corresponding to ABCB8(ATP-binding cassette, sub-family B (MDR/TAP), member 8) The peptide sequence was selected from the middle region of ABCB8. Peptide sequence EPVLFGTTIMENIRFGKLEASDEEVYTAAREANAHEFITSFPEGYNTVVG. | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Human: 100%; Mouse: 100%; Canine: 92%; Rabbit: 92%; Rat: 92%. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction