missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABCB8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
466.00€
Specifications
| Antigen | ABCB8 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
ABCB8 Polyclonal specifically detects ABCB8 in Human samples. It is validated for Western Blot.Specifications
| ABCB8 | |
| Polyclonal | |
| Rabbit | |
| Q9NUT2 | |
| 11194 | |
| Synthetic peptides corresponding to ABCB8(ATP-binding cassette, sub-family B (MDR/TAP), member 8) The peptide sequence was selected from the middle region of ABCB8. Peptide sequence EPVLFGTTIMENIRFGKLEASDEEVYTAAREANAHEFITSFPEGYNTVVG. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| ATP-binding cassette, sub-family B (MDR/TAP), member 8, EC 3.6.3, EST328128, M-ABC1mitochondrial ABC protein, mitochondrial, Mitochondrial ATP-binding cassette 1 | |
| ABCB8 | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title