Learn More
ABCC5 Mouse anti-Human, Unconjugated, Clone: 2H10F11, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67955-1-IG-150UL
Description
The ABCC5 antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human ABCC5. This antibody recognizes human, rat antigen. The ABCC5 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| ABCC5 | |
| Monoclonal | |
| Unconjugated | |
| 150 μL | |
| 10057 | |
| IgG1 |
| Western Blot | |
| 2H10F11 | |
| Mouse | |
| Primary | |
| Human, Rat |