missing translation for 'onlineSavingsMsg'
Learn More

ABCC5 Mouse anti-Human, Unconjugated, Clone: 2H10F11, Proteintech

Mouse Monoclonal Antibody

145.00€ - 410.00€

Specifications

Antigen ABCC5
Clone 2H10F11
Applications Western Blot
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17521466
View Documents
Proteintech
67955-1-IG-20UL
20 μL
145.00€
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17511466
View Documents
Proteintech
67955-1-IG-150UL
150 μL
410.00€
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The ABCC5 antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human ABCC5. This antibody recognizes human, rat antigen. The ABCC5 antibody has been validated for the following applications: WB, ELISA analysis.

Specifications

Specifications

ABCC5
Western Blot
Unconjugated
Human, Rat
IgG1
2H10F11
Monoclonal
Mouse
10057
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.