missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIMM44 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94065-0.1ml
This item is not returnable.
View return policy
Description
TIMM44 Polyclonal antibody specifically detects TIMM44 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| TIMM44 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| MIMT44, mitochondrial import inner membrane translocase subunit TIM44, mitochondrial inner membrane translocase, TIM44DKFZp686H05241, translocase of inner mitochondrial membrane 44 homolog (yeast) | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 153-452 of human TIMM44 (NP_006342.2). AKTAKQSAESVSKGGEKLGRTAAFRALSQGVESVKKEIDDSVLGQTGPYRRPQRLRKRTEFAGDKFKEEKVFEPNEEALGVVLHKDSKWYQQWKDFKENNVVFNRFFEMKMKYDESDNAFIRASRALTDKVTDLLGGLFSKTEMSEVLTEILRVDPAFDKDRFLKQCENDIIPNVLEAMISGELDILKDWCYEATYSQLAHPIQQAKALGLQFHSRILDIDNVDLAMGKMMEQGPVLIITFQAQLVMVVRNPKGEVVEGDPDKVLRMLYVWALCRDQDELNPYAAWRLLDISASSTEQIL | |
| 0.1 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 10469 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction