missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ACBD6 Monoclonal antibody specifically detects ACBD6 in Human samples. It is validated for Western Blot, ELISA, ELISA, ELISA
Specifications
Specifications
| Antigen | ACBD6 |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Clone | 4A9D5E9 |
| Conjugate | Janelia Fluor 549 |
| Dilution | Western Blot, ELISA, Sandwich ELISA, Sandwich ELISA Detection |
| Formulation | 50mM Sodium Borate |
| Gene Alias | acyl-CoA binding domain containing 6, acyl-CoA-binding domain-containing protein 6, acyl-Coenzyme A binding domain containing 6, MGC2404 |
| Host Species | Mouse |
| Immunogen | This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human ACBD6 (NP_115736.1; Met 1-Ala 282). |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?