missing translation for 'onlineSavingsMsg'
Learn More

Acetylthiocholine Iodide, TRC

Product Code. 30132872
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132872 25 g 25g
30134680 1 g 1g
30005584 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132872 Supplier Toronto Research Chemicals Supplier No. TRCA16461025G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1866-15-5
Molecular Formula C7 H16 N O S . I
Molecular Weight (g/mol) 289.1775
Synonym (2-Mercaptoethyl)trimethylammonium Acetate Iodide;
IUPAC Name 2-acetylsulfanylethyl(trimethyl)azanium;iodide
SMILES [I-].CC(=O)SCC[N+](C)(C)C

Specifications

Quantity 25 g
InChI Formula InChI=1S/C7H16NOS.HI/c1-7(9)10-6-5-8(2,3)4;/h5-6H2,1-4H3;1H/q+1;/p-1
Formula Weight 288.9997
Chemical Name or Material Acetylthiocholine Iodide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.