missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ACOT1 Polyclonal Antibody

Product Code. 15718311 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15718311 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15718311 Supplier Invitrogen™ Supplier No. PA560475

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: YKGETLPPVG VNRNRIKVTK DGYADIVDVL NSPLEGPDQK SFIPVERAES TFLFLVGQDD HNWK Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 70%.

Acyl-CoA thioesterases are a group of enzymes that catalyze the hydrolysis of acyl-CoAs to the free fatty acid and coenzyme A (CoASH), providing the potential to regulate intracellular levels of acyl-CoAs, free fatty acids and CoASH. Active towards fatty acyl-CoA with chain-lengths of C12-C16. Hunt M.C., Submitted (JUN-2005) to the EMBL/GenBank/DDBJ databases. Li W.B., Submitted (JAN-2003) to the EMBL/GenBank/DDBJ databases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACOT1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ACOT1
Gene Accession No. Q86TX2
Gene Alias ACH2; ACOT1; Acyl-CoA thioesterase 1; acyl-coenzyme A thioesterase 1; CTE1; CTE-1; CTE-I; CTE-Ib; Inducible cytosolic acyl-coenzyme A thioester hydrolase; L CTE-I; LACH2; Long chain acyl-CoA hydrolase; long chain acyl-CoA thioester hydrolase; Palmitoyl-coenzyme A thioesterase
Gene Symbols ACOT1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ACOT1. Recombinant protein control fragment (Product #RP-104360).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 641371
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.