missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adamantane-d16, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | Adamantane-d16 |
| CAS | 30470-60-1 |
| Molecular Formula | C10 D16 |
| Quantity | 0.25 g |
| Synonym | Tricyclo[3.3.1.13,7 ]decane-1,2,2,3,4,4,5,6,6,7,8,8,9,9,10,10-d16 (ACI), Adamantane-d16 (8 CI), Tricyclo[3.3.1.13,7 ]decane-d16 (9 CI), 1,2,2,3,4,4,5,6,6,7,8,8,9,9,10,10-Hexadecadeuterioadamantane, Perdeuteroadamantane, Adamantane-D16, Tricyclo[3.3.1.13,·]decane-d16 |
| InChI Formula | InChI=1 S/C10H16/c1-7-2-9-4-8(1)5-10(3-7)6-9/h7-10 H,1-6H2/i1D2,2D2,3D2,4D2,5D2,6D2,7 D,8 D,9 D,10 D |
| SMILES | [2 H]C1([2 H])C2([2 H])C([2 H])([2 H])C3([2 H])C([2 H])([2 H])C([2 H])(C([2 H])([2 H])C1([2 H])C3([2 H])[2 H])C2([2 H])[2 H] |
| IUPAC Name | 1,2,2,3,4,4,5,6,6,7,8,8,9,9,10,10-hexadecadeuterioadamantane |
| Molecular Weight (g/mol) | 152.333 |
| Formula Weight | 152.226 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction