missing translation for 'onlineSavingsMsg'
Learn More

Adrenalone, TRC

Product Code. 30004527
Change view
Click to view available options
Quantity:
100 mg
1 g
10 g
Unit Size:
100mg
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30004527 10 g 10g
30006158 1 g 1g
30011514 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30004527 Supplier Toronto Research Chemicals Supplier No. TRCA30509510G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-45-6
Molecular Formula C9 H11 N O3
Molecular Weight (g/mol) 181.19
Synonym Ethanone, 1-(3,4-dihydroxyphenyl)-2-(methylamino)-, Acetophenone, 3',4'-dihydroxy-2-(methylamino)- (7CI,8CI), Adrenalone (6CI), 1-(3,4-Dihydroxyphenyl)-2-(methylamino)ethanone, 3,4-Dihydroxy-α-methylaminoacetophenone, 3',4'-Dihydroxy-2-(methylamino)acetophenone, 4-Methylaminoacetopyrocatechol, Adrenolone, Adrenon, Adrenone, Chemosan, Haemodan, Kephrine, Ketogaze, Methaminoacetocatechol, NSC 243611, Stryphnon, Stryphnone, Styphnone, Stypnon, Stypnone, U 2134
IUPAC Name 1-(3,4-dihydroxyphenyl)-2-(methylamino)ethanone
SMILES CNCC(=O)c1ccc(O)c(O)c1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C9H11NO3/c1-10-5-9(13)6-2-3-7(11)8(12)4-6/h2-4,10-12H,5H2,1H3
Formula Weight 181.0739
Percent Purity >95%
Chemical Name or Material Adrenalone
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.