missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Adrenomedullin Monoclonal Antibody (C1), Invitrogen™
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA543983

 View more versions of this product

Product Code. 17674436

  • 453.00€ / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Adrenomedullin (ADM), a vasodilator produced by most contractile cells, is characterized by persistent hypotensive activity. ADM is involved in the regulation of fluid and electrolyte homeostasis and in the maintenance of cardiovascular functioning. In hypertensive patients, the level of ADM in plasma is upregulated. Natriuresis is a common systemic manifestation of aneurysmal subarachnoid hemorrhage. ADM has strong natriuretic actions. ADM-induced natriuresis is caused by an increase in glomerular filtration rate and a decrease in distal tubular sodium reabsorption. ADM is present both in the periphery and brain, and can exert central effects such as decreasing food ingestion.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Adrenomedullin
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
P35318
ADM
Ala22-Gly147 (Accession P35318), with N-terminal His Tag.
100 μL
Primary
Human
Antibody
IgG2a κ
Western Blot
C1
Unconjugated
ADM
ADM; Adrenomedullin; AM; AMpreproadrenomedullin; Ap; H39316; PAMP; preproadrenomedullin; proadrenomedullin N-20 terminal peptide; ProAM N-terminal 20 peptide; ProAM-N20; RATAP
Mouse
Protein A/G
RUO
133
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.