missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Adrenomedullin Monoclonal Antibody (C1), Invitrogen™

Mouse Monoclonal Antibody
180.00€ - 453.00€
Specifications
| Antigen | Adrenomedullin |
|---|---|
| Clone | C1 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17684436
|
Invitrogen™
MA543984 |
20 μL |
180.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17674436
![]() |
Invitrogen™
MA543983 |
100 μL |
453.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Adrenomedullin (ADM), a vasodilator produced by most contractile cells, is characterized by persistent hypotensive activity. ADM is involved in the regulation of fluid and electrolyte homeostasis and in the maintenance of cardiovascular functioning. In hypertensive patients, the level of ADM in plasma is upregulated. Natriuresis is a common systemic manifestation of aneurysmal subarachnoid hemorrhage. ADM has strong natriuretic actions. ADM-induced natriuresis is caused by an increase in glomerular filtration rate and a decrease in distal tubular sodium reabsorption. ADM is present both in the periphery and brain, and can exert central effects such as decreasing food ingestion.Specifications
| Adrenomedullin | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| ADM; Adrenomedullin; AM; AMpreproadrenomedullin; Ap; H39316; PAMP; preproadrenomedullin; proadrenomedullin N-20 terminal peptide; ProAM N-terminal 20 peptide; ProAM-N20; RATAP | |
| ADM | |
| IgG2a κ | |
| Protein A/G | |
| Antibody |
| C1 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| P35318 | |
| 133 | |
| Ala22-Gly147 (Accession P35318), with N-terminal His Tag. | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| ADM |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title