missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ AFAP Polyclonal Antibody
GREENER_CHOICE

Product Code. 15951385
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15951385 100 μg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15951385 Supplier Invitrogen™ Supplier No. PA572726

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

AFAP Polyclonal Antibody for Western Blot, ELISA

The protein encoded by this gene is a Src binding partner. It may represent a potential modulator of actin filament integrity in response to cellular signals, and may function as an adaptor protein by linking Src family members and/or other signaling proteins to actin filaments. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AFAP
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene Afap1
Gene Accession No. Q80YS6, Q8N556
Gene Alias 110 kDa actin filament-associated protein; 2600003E23Rik; 9630044L16Rik; actin filament associated protein 1; actin filament-associated protein 1; actin filament-associated protein, 110 kDa; AFAP; Afap1; AFAP110; AFAP-110; AI848729; FLJ56849; im:7148276; Kiaa3018; mKIAA3018
Gene Symbols Afap1
Host Species Rabbit
Immunogen AFAP1 antibody for an 18 amino acid synthetic peptide near the amino terminus of human AFAP1. The immunogen is within amino acids 30 - 80 of AFAP1.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60312, 70292
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.