missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aflatoxin B1 Antibody (6A10) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 21 publication
Brand: Novus Biologicals NB600-443SS
This item is not returnable.
View return policy
Description
Aflatoxin B1 Monoclonal specifically detects Aflatoxin B1 in Human, All Species samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Dot Blot.
Specifications
| Aflatoxin B1 | |
| Monoclonal | |
| 1.0 mg/ml | |
| Western Blot 1:500-1:2000, ELISA 5 ug/ml, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100. Use reported in scientific literature (PMID 24114584), Immunohistochemistry-Paraffin reported in scientific literature (PMID 24391771), Immunohistochemistry-Frozen, Dot Blot reported in scientific literature (PMID 19524575) | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, All species | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 6A10 | |
| Unconjugated | |
| AFB1, AFB1-AR1, Aldoketoreductase 7 | |
| Aflatoxin B1 DNA (against the midazole ring-opened persistent form of the major N-7 guanine adduct of AFB1). | |
| 0.025 mL | |
| Cancer | |
| All species. There is no cross-reactivity with other carcinogen-DNA adducts. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction