missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AK6 Monoclonal Antibody (B1-F4)

Description
AK6 Monoclonal Antibody for ICC/IF, Western Blot
Broad-specificity nucleoside monophosphate (NMP) kinase that catalyzes the reversible transfer of the terminal phosphate group between nucleoside triphosphates and monophosphates. AMP and dAMP are the preferred substrates, but CMP and dCMP are also good substrates. IMP is phosphorylated to a much lesser extent. All nucleoside triphosphates ATP, GTP, UTP, CTP, dATP, dCTP, dGTP, and TTP are accepted as phosphate donors. CTP is the best phosphate donor, followed by UTP, ATP, GTP and dCTP. May have a role in nuclear energy homeostasis. Has also ATPase activity. May be involved in regulation of Cajal body (CB) formation.
Specifications
Specifications
| Antigen | AK6 |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence, Western Blot |
| Classification | Monoclonal |
| Clone | B1-F4 |
| Concentration | 2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% Glycerol, 0.2% BSA and 0.05% sodium azide; pH 7.4 |
| Gene | AK6 |
| Gene Accession No. | Q5EB68, Q8VCP8, Q9Y3D8 |
| Gene Alias | 2810046E22Rik; 4921516M08Rik; AD-004; adenylate kinase 6; adenylate kinase isoenzyme 6; adenylate kinase isoenzyme 6 {ECO:0000255; Adrenal gland protein AD-004; AK/ATPase; AK/ATPase {ECO:0000255; AK6; AK6 {ECO:0000255; ATP-AMP transphosphorylase 6; CGI-137; CINAP; CIP; coilin interacting nuclear ATPase protein; coilin interacting protein; coilin-interacting nuclear ATPase protein; coilin-interacting nuclear ATPase protein {ECO:0000255; Dual activity adenylate kinase/ATPase; dual activity adenylate kinase/ATPase {ECO:0000255; HAMAP-Rule:MF_03173}; hCINAP |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?