missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Antibody detects endogenous levels of total AKAP2.
Binds to regulatory subunit (RII) of protein kinase A. May be involved in establishing polarity in signaling systems or in integrating PKA-RII isoforms with downstream effectors to capture, amplify and focus diffuse, trans-cellular signals carried by cAMP. [UniProt].
Specifications
Specifications
| Antigen | AKAP2 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | AKAP2 |
| Gene Accession No. | Q9Y2D5 |
| Gene Alias | A kinase (PRKA) anchor protein 2; AA959716; AI649048; AKAP expressed in kidney and lung; AKAP2; AKAP-2; AKAPKL; AKAP-KL; A-kinase anchor protein 2; A-kinase anchoring protein 2; B230340M18Rik; KL2B; MISP2; PALM2-AKAP2; PRKA2; protein kinase A anchoring protein 2; protein kinase A2; protein kinase A-anchoring protein 2 |
| Gene Symbols | AKAP2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
