missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKAP5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-54807
This item is not returnable.
View return policy
Description
AKAP5 Polyclonal specifically detects AKAP5 in Human samples. It is validated for Western Blot.
Specifications
| AKAP5 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| A kinase (PRKA) anchor protein 5, AKAP 79, AKAP7979kDa, A-kinase anchor protein 79 kDa, A-kinase anchoring protein 75/79, cAMP-dependent protein kinase regulatory subunit II high affinity bindingprotein, cAMP-dependent protein kinase regulatory subunit II high affinity-bindingprotein, H21 | |
| Rabbit | |
| 47 kDa | |
| 100 μL | |
| Primary | |
| Porcine: 86%; Equine: 86%; Yeast: 82%; . | |
| Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Yeast | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| P24588 | |
| AKAP5 | |
| Synthetic peptides corresponding to AKAP5(A kinase (PRKA) anchor protein 5) The peptide sequence was selected from the middle region of AKAP5 (NP_004848). Peptide sequence KQFLISAENEQVGVFANDNGFEDRTSEQYETLLIETASSLVKNAIQLSIE. | |
| Affinity purified | |
| RUO | |
| 9495 | |
| Centrifuge vial prior to reconstitution. Add 50μL distilled water to a final antibody concentration of 1mg/mL. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction