missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
AKT Monoclonal antibody specifically detects AKT in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | AKT |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 0002 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:200-1:1000 |
| Formulation | 0.2 um filtered solution in PBS |
| Gene Alias | RAC-ALPHA, AKT, AKT serine/threonine kinase 1, PKB, PKB-ALPHA, PRKBA, RAC |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with a synthetic peptide corresponding to the residues around Ser308 of Human phospho-AKT. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?