missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aldosterone Antibody (BGN/10/42110) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-62313
This item is not returnable.
View return policy
Description
Aldosterone Monoclonal specifically detects Aldosterone in All Species samples. It is validated for ELISA.
Specifications
| Aldosterone | |
| Monoclonal | |
| 1.0 mg/mL | |
| ELISA 1:100-1:2000 | |
| Aldosterone (3-CMO) conjugated to Bovine Serum Albumin. | |
| 0.1 mg | |
| Lipid and Metabolism | |
| This product is specific for aldosterone and exhibits minimal reactivity with androstenedione, corticosterone, DHEA and desoxycortisol. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| ELISA | |
| BGN/10/42110 | |
| Unconjugated | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| All species | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction