missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aldosterone Antibody (BGN/10/42110) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
190.00€ - 487.00€
Specifications
| Antigen | Aldosterone |
|---|---|
| Clone | BGN/10/42110 |
| Concentration | 1.0 mg/mL |
| Dilution | ELISA 1:100-1:2000 |
| Applications | ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18052356
|
Novus Biologicals
NB100-62313-0.025MG |
0.025 mg |
190.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18798293
|
Novus Biologicals
NB100-62313 |
0.1 mg |
487.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Aldosterone Monoclonal specifically detects Aldosterone in All Species samples. It is validated for ELISA.Specifications
| Aldosterone | |
| 1.0 mg/mL | |
| ELISA | |
| Unconjugated | |
| RUO | |
| All species | |
| IgG1 | |
| Affinity Purified | |
| This product is specific for aldosterone and exhibits minimal reactivity with androstenedione, corticosterone, DHEA and desoxycortisol. |
| BGN/10/42110 | |
| ELISA 1:100-1:2000 | |
| Monoclonal | |
| Mouse | |
| Lipid and Metabolism | |
| Aldosterone (3-CMO) conjugated to Bovine Serum Albumin. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title