missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Allophycocyanin Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB8927
This item is not returnable.
View return policy
Description
Allophycocyanin Monoclonal specifically detects Allophycocyanin in Multi-Species samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications
| Allophycocyanin | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| APC | |
| Allophycocyanin isolated from red algae Accession # P25054 | |
| 100 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Flow Cytometry, CyTOF | |
| 936809 | |
| Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| P25054 | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Detects Allophycocyanin in flow cytometry. | |
| Multi-species | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction