missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Allophycocyanin Antibody, R&D Systems™

Mouse Monoclonal Antibody
209.00€ - 420.00€
Specifications
| Antigen | Allophycocyanin |
|---|---|
| Clone | 936809 |
| Dilution | Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready |
| Applications | Flow Cytometry, CyTOF |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18776048
![]() |
R&D Systems
MAB8927-SP |
25 μg |
209.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18786078
![]() |
R&D Systems
MAB8927 |
100 μg |
420.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Allophycocyanin Monoclonal specifically detects Allophycocyanin in Multi-Species samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
| Allophycocyanin | |
| Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| APC | |
| IgG2b | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 936809 | |
| Flow Cytometry, CyTOF | |
| Unconjugated | |
| Mouse | |
| Multi-species | |
| P25054 | |
| Allophycocyanin isolated from red algae Accession # P25054 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects Allophycocyanin in flow cytometry. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title