Learn More
alpha 1-Acid Glycoprotein Antibody (SR1818), R&D Systems™
Product Code. 18601985 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
Unit Size:
100µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18601985 100 μg 100µL
18633614 50 μg 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 18601985 Supplier Novus Biologicals Supplier No. NBP321776100ul
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
A Novus product, part of R&D Systems. Rabbit Monoclonal Antibody
alpha 1-Acid Glycoprotein Monoclonal specifically detects alpha 1-Acid Glycoprotein in Human samples. It is validated for Immunohistochemistry∣Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Antigen alpha 1-Acid Glycoprotein
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone SR1818
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200
Formulation PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Gene Alias AGP 1, AGP1alpha-1-acid glycoprotein 1, AGP-A, OMD 1, ORM, orosomucoid 1, orosomucoid-1
Host Species Rabbit
Immunogen A synthesized peptide derived from human alpha 1-Acid Glycoprotein (Uniprot #: P02763)
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 5004
Target Species Human
Content And Storage Store at -20° C. Avoid freeze/thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus