missing translation for 'onlineSavingsMsg'
Learn More

alpha,alpha,alpha-Trifluoro-m-tolyl isocyanate, 98%, ACROS Organics™

Product Code. 10424253 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10424253 5 g Glass bottle 5g
10587933 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10424253 Supplier Thermo Scientific Acros Supplier No. 170350050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 329-01-1
Molecular Formula C8H4F3NO
Molecular Weight (g/mol) 187.12
MDL Number MFCD00002020
InChI Key SXJYSIBLFGQAND-UHFFFAOYSA-N
Synonym 3-trifluoromethyl phenyl isocyanate, 1-isocyanato-3-trifluoromethyl benzene, benzene, 1-isocyanato-3-trifluoromethyl, a,a,a-trifluoro-m-tolyl isocyanate, isocyanobenzotrifluoride, unii-qps7llz6xm, 3-trifluoromethylphenyl isocyanate, alpha,alpha,alpha-trifluoro-m-tolyl isocyanate, 3-trifluoromethyl phenylisocyanate, qps7llz6xm
PubChem CID 9483
IUPAC Name 1-isocyanato-3-(trifluoromethyl)benzene
SMILES FC(F)(F)C1=CC=CC(=C1)N=C=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 1.3360g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula CF3C6H4NCO
Refractive Index 1.4670 to 1.4690 (20°C, 589nm)
Quantity 5 g
Formula Weight 187.12
Percent Purity ≥97.5%
Physical Form Liquid
Chemical Name or Material α, α, α-Trifluoro-m-tolyl isocyanate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.