missing translation for 'onlineSavingsMsg'
Learn More

Alpha-Descyclohexyl-Alpha-phenyl Oxybutynin, TRC

Product Code. 30357900
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30357900 10 mg 10mg
30367273 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30357900 Supplier Toronto Research Chemicals Supplier No. TRCD28884010MG
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14943-53-4
Molecular Formula C22 H25 N O3
Molecular Weight (g/mol) 351.44
Synonym 4-(Diethylamino)but-2-ynyl 2-Hydroxy-2,2-diphenylacetate, Oxybutynin Hydrochloride Imp. B (EP), Diphenyl Analogue of Oxybutynin
IUPAC Name 4-(diethylamino)but-2-ynyl 2-hydroxy-2,2-diphenylacetate
SMILES CCN(CC)CC#CCOC(=O)C(O)(c1ccccc1)c2ccccc2

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C22H25NO3/c1-3-23(4-2)17-11-12-18-26-21(24)22(25,19-13-7-5-8-14-19)20-15-9-6-10-16-20/h5-10,13-16,25H,3-4,17-18H2,1-2H3
Formula Weight 351.1834
Chemical Name or Material 4-(Diethylamino)but-2-ynyl 2-Hydroxy-2,2-diphenylacetate (Diphenyl Analogue of Oxybutynin)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.