Learn More
Brady™ Aluminum Warning Sign with Polyester Overlaminate
5.55€ - 27.05€
Specifications
Certifications/Compliance | ISO 7010 |
---|---|
Material | Aluminum, Polyester |
Quantity | 1 Ea. |
Description | Aluminum with Polyester Overlaminate |
For Use With (Application) | Indoor and outdoor safety signs |
Product Code | Brand | Height (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Height (Metric) | Price | Quantity & Availability | |||||
11784854
|
Brady™
255201 |
315 mm |
27.05€
Each |
Estimated Shipment: 14-06-2024
Log in to see stock. |
|||||
11886341
|
Brady™
255200 |
200 mm |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
11710294
|
Brady™
241644 |
100 mm |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
Specifications
ISO 7010 | |
1 Ea. | |
Indoor and outdoor safety signs | |
-40°C to 140°C | |
Warning of Flammable Materials | |
Black on Yellow |
Aluminum, Polyester | |
Aluminum with Polyester Overlaminate | |
0.271 mm | |
Triangular | |
Warning Sign |