missing translation for 'onlineSavingsMsg'
Learn More

Amino Albendazole Sulfone, TRC

Product Code. 30133241
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133241 50 mg 50mg
30138374 100 mg 100mg
18042420 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133241 Supplier Toronto Research Chemicals Supplier No. TRCA58095050MG
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80983-34-2
Molecular Formula C10 H13 N3 O2 S
Molecular Weight (g/mol) 239.29
Synonym 1H-Benzimidazol-2-amine, 5-(propylsulfonyl)- (9CI), 6-(Propylsulfonyl)-1H-benzimidazol-2-amine, 2-Amino-5-1H-propylsulfonylbenzimidazole, 2-Aminoalbendazole sulfone, 5-Propylsulfonyl-1H-benzimidazol-2-amine, Albendazole-2-aminosulfone, SKF 81038
IUPAC Name 5-propylsulfonyl-1H-benzimidazol-2-amine
SMILES CCCS(=O)(=O)c1ccc2[nH]c(N)nc2c1

Specifications

Analyte or Component Names Albendazole-2-aminosulfone
Shipping Condition Room Temperature
InChI Formula InChI=1S/C10H13N3O2S/c1-2-5-16(14,15)7-3-4-8-9(6-7)13-10(11)12-8/h3-4,6H,2,5H2,1H3,(H3,11,12,13)
Solution Type Neat
Industry Type Pharmaceutical
Quantity 50 mg
Formula Weight 239.0728
Percent Purity >95%
Chemical Name or Material Albendazole-2-aminosulfone
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.