missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Androgen Receptor Polyclonal Antibody
GREENER_CHOICE

Product Code. 11565922
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11565922 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11565922 Supplier Invitrogen™ Supplier No. PA1111B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PA1-111B detects androgen receptor (AR) from human samples. This antibody does not detect estrogen, progesterone, or glucocorticoid receptors. PA1-111B has been successfully used in Western blot procedures. By Western blot, this antibody detects an ~110 kDa protein representing AR from HepG2 nuclear extracts. The PA1-111B immunogen is a synthetic peptide corresponding to residues M(1) E V Q L G L G R V Y P R P P S K T Y R G(21) C of human AR. This sequence is completely conserved between human, mouse, and rat AR. The PA1-111B immunizing peptide (Cat. # PEP-006) is available for use in neutralization and control experiments.

The androgen receptor (AR) is a member of the steroid-hormone receptor superfamily of nuclear receptors. The receptor is more than 90 kDa and has three major functional domains: the N-terminal domain, DNA-binding domain, and the androgen-binding domain. The androgen receptor is a ligand-activated transcription factor that binds active testosterone (T) and dihydrotestosterone (DHT). Upon binding the hormone ligand, the receptor dissociates from accessory proteins, translocates into the nucleus, dimerizes, and then stimulates transcription of androgen responsive genes. The AR signaling pathway plays a key role in development and function of male reproductive organs, including the prostate and epididymis.AR also plays a role in nonreproductive organs, such as muscle, hair follicles, and brain.Androgen Receptor is a phosphoprotein, and also regulates mitogen-activated protein kinase (MAP kinase). The inhibition of the MEK1/2 pathway correlates directly with a change in phosphorylation state of the androgen receptor. Abnormalities in the AR signaling pathway have been linked to a number of diseases, including prostate cancer, Kennedy's disease, and male infertility. Mutations in this gene are associated with complete androgen insensitivity (CAIS).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Androgen Receptor
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1 mg/mL BSA and 0.05% sodium azide
Gene AR
Gene Accession No. P10275
Gene Alias AIS; Andr; androgen nuclear receptor variant 2; androgen receptor; androgen receptor (Testicular feminization), same as Tfm; androgen receptor splice variant 4b; Ar; AR8; AR-A; AW320017; DHTR; dihydrotestosterone receptor; HUMARA; HYSP1; KD; NR3C4; Nuclear receptor subfamily 3 group C member 4; RP11-383C12.1; SBMA; SMAX1; TFM; Variant AR45
Gene Symbols AR
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues M(1) E V Q L G L G R V Y P R P P S K T Y R G(21) C of human AR.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 367
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.