missing translation for 'onlineSavingsMsg'
Learn More

Angiopoietin-like Protein 1/ANGPTL1 Antibody (3A5), Novus Biologicals™

Product Code. 18319568 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18319568 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18319568 Supplier Novus Biologicals Supplier No. H00009068M03

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Angiopoietin-like Protein 1/ANGPTL1 Monoclonal antibody specifically detects Angiopoietin-like Protein 1/ANGPTL1 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Angiopoietin-like Protein 1/ANGPTL1
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 3A5
Conjugate Unconjugated
Dilution ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH50640
Gene Alias ANG-3, ANG3angioarrestin, angiopoietin Y1, angiopoietin-3, angiopoietin-like 1, Angiopoietin-like protein 1, angiopoietin-related protein 1, ANGPT3, AngY, ARP1angiopoietin 3, dJ595C2.2, dJ595C2.2 (angiopoietin Y1), KIAA0351, UNQ162
Host Species Mouse
Immunogen ANGPTL1 (AAH50640, 24 a.a. ~ 491 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. GQFKIKKINQRRYPRATDGKEEAKKCAYTFLVPEQRITGPICVNTKGQDASTIKDMITRMDLENLKDVLSRQKREIDVLQLVVDVDGNIVNEVKLLRKESRNMNSRVTQLYMQLLHEIIRKRDNSLELSQLENKILNVTTEMLKMATRYRELEVKYASLTDLVNNQSVMITLLEEQCLRIFSRQDTHVSPPLVQVVPQHIPNSQQYTPGLLGGNEIQRDPGYPRDLMPPPDLATSPTKSPFKIPPVTFINEGPFKDCQQAKEAGHSVSGIYMIKPENSNGPMQLWCENSLDPGGWTVIQKRTDGSVNFFRNWENYKKGFGNIDGEYWLGLENIYMLSNQDNYKLLIELEDWSDKKVYAEYSSFRLEPESEFYRLRLGTYQGNAGDSMMWHNGKQFTTLDRDKDMYAGNCAHFHKGGWWYNACAHSNLNGVWYRGGHYRSKHQDGIFWAEYRGGSYSLRAVQMMIKPID
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9068
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.