missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ANKK1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16687684
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16687684 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16687684 Supplier Invitrogen™ Supplier No. PA5101209

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ANKK1.

ANKK1 serine/threonine kinase is a member of the RIPK family and is closely linked to the DRD2 gene. ANKK1 is closely linked to DRD2 gene on chr 11, and a well studied restriction fragment length polymorphism designated TaqIA, was originally associated with the DRD2 gene, however, later was determined to be located in exon 8 of ANKK1 gene, where it causes a nonconservative amino acid substitution. It is not clear if ANKK1 plays any role in neuropsychiatric disorders previously associated with Taq1A RFLP.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANKK1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ANKK1
Gene Accession No. Q8NFD2
Gene Alias 9930020N01Rik; ANKK1; ankyrin repeat and kinase domain containing 1; ankyrin repeat and protein kinase domain-containing protein 1; PKK2; Protein kinase PKK2; sgK288; Sugen kinase 288; X-kinase
Gene Symbols ANKK1
Host Species Rabbit
Immunogen A synthesized peptide derived from human ANKK1(Accession Q8NFD2), corresponding to amino acid residues W462-L512.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 255239
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.