missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ANKRD10 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17161274 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17161274 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17161274 Supplier Invitrogen™ Supplier No. PA5113415

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ANKRD10 Polyclonal Antibody for Western Blot, ELISA

ANKRD10 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANKRD10
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene ANKRD10
Gene Accession No. Q99LW0, Q9NXR5
Gene Alias 4833425P12Rik; ANKRD10; ankyrin repeat domain 10; ankyrin repeat domain-containing protein 10; AW549277
Gene Symbols ANKRD10
Host Species Rabbit
Immunogen Recombinant Human Ankyrin repeat domain-containing protein 10 protein (1-220AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 102334, 55608
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.