Learn More
Invitrogen™ Annexin A6 Recombinant Rabbit Monoclonal Antibody (7X4H2)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA558020
Description
Immunogen sequence: DDDAAGQFF PEAAQVAYQM WELSAVARVE LKGTVRPAND FNPDADAKAL RKAMKGLGTD EDTIIDIITH RSNVQRQQIR QTFKSHFGRD LMTDLKSEIS GDLARLILGL MMPPAHYDAK QLKKAMEGAG TDEKALIEIL ATRTNAEIRA INEAYKEDYH KSLEDALSSD TSGHFRRILI SLATGHREEG GENLDQARED AQVAAEILEI ADTPSGDKTS LETRFMTILC TRSYPHLRRV FQEFIKMTNY DVEHTIKKEM SGDVRDAFVA IVQSVKNKPL FFADKLYKSM KGAGTDEKTL TRIMVSRSEI DLLNIRREFI EKYDKSLHQA IEGDTSGDFL KALLALCGGE D.
Annexin VI belongs to a family of calcium-dependent membrane and phospholipid binding proteins. Although their functions are still not clearly defined, several members of the annexin family have been implicated in membrane-related events along exocytotic and endocytotic pathways. The annexin VI gene is approximately 60 kbp long and contains 26 exons. It encodes a protein of about 68 kDa that consists of eight 68-amino acid repeats separated by linking sequences of variable lengths. It is highly similar to human annexins I and II sequences, each of which contain four such repeats. Exon 21 of annexin VI is alternatively spliced, giving rise to two isoforms that differ by a 6-amino acid insertion at the start of the seventh repeat. Annexin VI has been implicated in mediating the endosome aggregation and vesicle fusion in secreting epithelia during exocytosis.
Specifications
| Annexin A6 | |
| Recombinant Monoclonal | |
| 0.71 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| P08133, P14824, P48037 | |
| ANXA6 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 324-673 of human Annexin A6/Annexin VI (NP_001146.2). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 7X4H2 | |
| Unconjugated | |
| ANXA6 | |
| 67 kDa calelectrin; Annexin A6; annexin VI; annexin VI (p68); annexin VI, p68; annexin-6; Anx6; ANXA6; AnxVI; AW107198; Cabm; calcium-binding protein 65/67; calcium-binding protein CATA 65/67; calcium-binding protein p68; calelectrin; calphobindin II; calphobindin-II; Camb; CBP 65/67; CBP68; chromobindin-20; CPB-II; lipocortin VI; p68; p70; Protein III; testis secretory sperm-binding protein Li 198a | |
| Rabbit | |
| Protein A | |
| RUO | |
| 11749, 309, 79125 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.