missing translation for 'onlineSavingsMsg'
Learn More

γ-Catenin Mouse, Unlabeled, Clone: 15, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 610253

 View more versions of this product

Product Code. 15895359

  • ONLINE EXCLUSIVE PRICE
    179.00€
    178.69€ / 1 set
    Save 0.31€
    0.17% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

γ-Catenin (plakoglobin) was identified as a compenent of desmosomes where it associates with desmoglein. γ-Catenin and β-Catenin are closely related proteins that have significant homology with the Drosophila armadillo protein. In addition to complexing with E-Cadherin, γ-Catenin and β-Catenin have been observed in association with the intracellular domain of N-Cadherin. It has been proposed that one molecule of α-Catenin and at least one molecule of β-Catenin and γ-Catenin simultaneously bind to a single cadherin molecule. A 19 amino acid sequence of desmoglein (Dsg1) was found to be critical for binding of γ-Catenin. This region has significant homology to the catenin-binding domain of classical cadherins, thus suggesting a common mechanism for γ-Catenin localization at both adherens junctions and desmosomes.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY
Specifications

Specifications

γ-Catenin
Monoclonal
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Human γ-Catenin aa. 553-738
50 μg
Cell Biology
Canine, Chicken, Human, Mouse, Rat
IgG2a
Western Blot
15
Unconjugated
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.