missing translation for 'onlineSavingsMsg'
Learn More

γ-Catenin Mouse, Unlabeled, Clone: 15, BD

Mouse Monoclonal Antibody

178.69€ - 323.14€

Specifications

Antigen γ-Catenin
Clone 15
Concentration 250μg/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15895359
View Documents
BD Biosciences
610253
50 μg
ONLINE EXCLUSIVE PRICE
179.00€
178.69€ / 1 set
Save 0.31€
0.17% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15805369
View Documents
BD Biosciences
610254
150 μg
ONLINE EXCLUSIVE PRICE
325.00€
323.14€ / 1 set
Save 1.86€
0.57% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

γ-Catenin (plakoglobin) was identified as a compenent of desmosomes where it associates with desmoglein. γ-Catenin and β-Catenin are closely related proteins that have significant homology with the Drosophila armadillo protein. In addition to complexing with E-Cadherin, γ-Catenin and β-Catenin have been observed in association with the intracellular domain of N-Cadherin. It has been proposed that one molecule of α-Catenin and at least one molecule of β-Catenin and γ-Catenin simultaneously bind to a single cadherin molecule. A 19 amino acid sequence of desmoglein (Dsg1) was found to be critical for binding of γ-Catenin. This region has significant homology to the catenin-binding domain of classical cadherins, thus suggesting a common mechanism for γ-Catenin localization at both adherens junctions and desmosomes.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

Specifications

Specifications

γ-Catenin
250μg/mL
Monoclonal
Mouse
Cell Biology
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
IgG2a
Affinity Purified
15
Western Blot
Unconjugated
RUO
Canine, Chicken, Human, Mouse, Rat
Human γ-Catenin aa. 553-738
Primary
Store undiluted at -20°C.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.