missing translation for 'onlineSavingsMsg'
Learn More

Anti-Human STRO-1 (STRO-1), Proteintech

Mouse Monoclonal Antibody

104.00€ - 320.00€

Specifications

Antigen STRO-1
Clone STRO-1
Concentration 0.5 mg/mL
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30234044
View Documents
Proteintech
65184-1-IG-500UG
500 μg
ONLINE EXCLUSIVE PRICE
322.08€
320.00€ / 500µg
Save 2.08€
0.65% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30236665
View Documents
Proteintech
65184-1-IG-100UG
100 μg
ONLINE EXCLUSIVE PRICE
104.68€
104.00€ / 100µg
Save 0.68€
0.65% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The STRO-1 antibody from Proteintech is a mouse monoclonal antibody generated with human cd34 positive bone marrow cells of human STRO-1. This antibody recognizes human antigen. The STRO-1 antibody has been validated for the following applications: FC analysis.
Specifications

Specifications

STRO-1
0.5 mg/mL
Monoclonal
Liquid
RUO
Human CD34 positive bone marrow cells
Primary
Store at 2-8°C. Stable for one year after shipment.
STRO-1
Flow Cytometry
Unconjugated
Mouse
Human
IgM λ
Purified by precipitation and chromatography
Flow Antibody
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.