missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anti-Mouse Ly-6G (1A8), Proteintech
Rat Monoclonal Antibody
Brand: Proteintech 65078-1-IG-500UG
This item is not returnable.
View return policy
Description
The Ly-6G antibody from Proteintech is a rat monoclonal antibody generated with ly-6g transfected el-4j cell line of mouse Ly-6G. This antibody recognizes mouse antigen. The Ly-6G antibody has been validated for the following applications: FC analysis.Specifications
| Ly-6G | |
| Monoclonal | |
| 500 μg/mL | |
| Rat | |
| Affinity purification | |
| RUO | |
| 546644 | |
| Store at 2-8°C. Stable for one year after shipment. | |
| Liquid |
| Flow Cytometry | |
| 1A8 | |
| Unconjugated | |
| Ly-6G transfected EL-4J cell line | |
| 500 μg | |
| Primary | |
| Mouse | |
| Flow Antibody | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction