missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anti-Mouse Ly-6G (1A8), Proteintech
Rat Monoclonal Antibody
37.00€ - 112.00€
Specifications
| Antigen | Ly-6G |
|---|---|
| Clone | 1A8 |
| Concentration | 500 μg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30238034
|
Proteintech
65078-1-IG-500UG |
500 μg |
112.73€ 112.00€ / 500µg Save 0.73€ 0.65% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30236559
|
Proteintech
65078-1-IG-100UG |
100 μg |
38.00€ 37.00€ / 100µg Save 1.00€ 2% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Ly-6G antibody from Proteintech is a rat monoclonal antibody generated with ly-6g transfected el-4j cell line of mouse Ly-6G. This antibody recognizes mouse antigen. The Ly-6G antibody has been validated for the following applications: FC analysis.Specifications
| Ly-6G | |
| 500 μg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 546644 | |
| IgG2a κ | |
| Affinity purification | |
| Flow Antibody |
| 1A8 | |
| Flow Cytometry | |
| Unconjugated | |
| Rat | |
| Mouse | |
| Ly-6G transfected EL-4J cell line | |
| Primary | |
| Store at 2-8°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title