missing translation for 'onlineSavingsMsg'
Learn More

Neuronal Pentraxin Mouse, Unlabeled, Clone: 22, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 610370

 View more versions of this product

Product Code. 15855419

  • ONLINE EXCLUSIVE PRICE
    325.00€
    323.14€ / 1 set
    Save 1.86€
    0.57% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Neuronal pentraxin (NP) is a potential receptor for the snake venom toxin, taipoxin. The highest levels of NP mRNA are found in the cerebellum and the CA3 region of the hippocampus. More specifically, NP mRNA is expressed at high levels in Purkinje and granule neurons of the cerebellum and CA3 neurons of the hippocampus. Moderate levels are present in neurons of the hilus, dentate gyrus, CA1, cerebral cortex, medial habenula, anterior hypothalamic nuclei, olfactory cortex, and inferior olivary nuclei. NP mRNA is not expressed in glia of white or gray matter. It is thought that NP serves as a receptor for taipoxin and, after binding, NP-taipoxin is taken into presynaptic terminals. Alternatively, the binding of taipoxin by NP may represent a nonspecific mechanism to sequester toxins from synapses.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Neuronal Pentraxin
Monoclonal
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
150 μg
Cell Biology
Human, Rat
IgG1
Western Blot
22
Unconjugated
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.