Learn More
Neuronal Pentraxin Mouse, Unlabeled, Clone: 22, BD
Mouse Monoclonal Antibody
175.52€ - 323.14€
Specifications
| Antigen | Neuronal Pentraxin |
|---|---|
| Clone | 22 |
| Concentration | 250μg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15845419
|
BD Biosciences
610369 |
50 μg |
176.00€ 175.52€ / 1 set Save 0.48€ 0.27% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15855419
|
BD Biosciences
610370 |
150 μg |
325.00€ 323.14€ / 1 set Save 1.86€ 0.57% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Neuronal pentraxin (NP) is a potential receptor for the snake venom toxin, taipoxin. The highest levels of NP mRNA are found in the cerebellum and the CA3 region of the hippocampus. More specifically, NP mRNA is expressed at high levels in Purkinje and granule neurons of the cerebellum and CA3 neurons of the hippocampus. Moderate levels are present in neurons of the hilus, dentate gyrus, CA1, cerebral cortex, medial habenula, anterior hypothalamic nuclei, olfactory cortex, and inferior olivary nuclei. NP mRNA is not expressed in glia of white or gray matter. It is thought that NP serves as a receptor for taipoxin and, after binding, NP-taipoxin is taken into presynaptic terminals. Alternatively, the binding of taipoxin by NP may represent a nonspecific mechanism to sequester toxins from synapses.
Specifications
| Neuronal Pentraxin | |
| 250μg/mL | |
| Monoclonal | |
| Mouse | |
| Cell Biology | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| IgG1 | |
| Affinity Purified |
| 22 | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human, Rat | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| Primary | |
| Store undiluted at -20°C. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.