Learn More
WIP (pS488) Mouse, PE, Clone: K32-824, BD
Description
Wiskott-Adrich syndrome protein (WASP)-Interacting Protein (WIP) is a member of the verprolin family of proteins that regulate cytoskeletal organization in a wide variety of cellular activities, including endocytosis, cellular adhesion and migration, mast cell degranulation, and lymphocyte activation. The 503-amino acid WIP protein contains binding sites for actin (globular and filamentous) and other proteins that are involved in the regulation of actin polymerization, such as WASP, N-WASP, profilin, cortactin, Hck, and NCK. As its functions imply, WIP is localized in actin-rich cell structures.
Intracellular Staining
Specifications
Specifications
| Antigen | WIP (pS488) |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | K32-824 |
| Conjugate | PE |
| Description | PRPL-2 protein, WAIP, WASIP, WASPIP |
| Formulation | Aqueous buffered solution containing BSA and ≤0.09% sodium azide. |
| Host Species | Mouse |
| Immunogen | Phosphorylated Human WIP Peptide |
| Purification Method | Affinity Purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.