missing translation for 'onlineSavingsMsg'
Learn More

AP2A2 Antibody, Alexa Fluor™ 750, Novus Biologicals™

Product Code. 18542903 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18542903 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18542903 Supplier Novus Biologicals Supplier No. NBP299239AF750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

AP2A2 Polyclonal antibody specifically detects AP2A2 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen AP2A2
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 750
Formulation 50mM Sodium Borate
Gene Alias 100 kDa coated vesicle protein C, Adapter-related protein complex 2 alpha-2 subunit, adaptin, alpha B, Adaptor protein complex AP-2 subunit alpha-2, adaptor-related protein complex 2, alpha 2 subunit, alpha2-adaptin, Alpha-adaptin C, alpha-adaptin C; Huntingtin interacting protein J, AP-2 complex subunit alpha-2, Clathrin assembly protein complex 2 alpha-C large chain, clathrin-associated/assembly/adaptor protein, large, alpha 2, DKFZP564D1864, HIP9ADTAB, Huntingtin yeast partner J, Huntingtin-interacting protein 9, Huntingtin-interacting protein J, HYPJCLAPA2, KIAA0899HIP-9, Plasma membrane adaptor HA2/AP2 adaptin alpha C subunit
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human AP2A2.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neurodegeneration, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 161
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.